ENA455585878 Enamine

3-(trifluoromethyl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD11100076
  • Cas Number : 1076245-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-195893
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart