ENA455585946 Enamine

2-chloro-4H,5H,6H-cyclopenta[b]thiophen-4-amine

  • MDL number MFCD24076085
  • Cas Number : 1518717-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-216513
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart