ENA455586020 Enamine

7-azabicyclo[2.2.1]heptan-2-ylmethanol hydrochloride

  • MDL number MFCD28714712
  • Cas Number : 1803570-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-216658
Availability details for
Quantity
  • Availability
  • Add to Cart