ENA455586043 Enamine

2,2-dimethyl-1-(pyrrolidin-1-yl)propan-1-one

  • MDL number MFCD01326710
  • Cas Number : 3389-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-216764
Availability details for
Quantity
  • Availability
  • Add to Cart