ENA455586076 Enamine

N-{[1-(2-chlorophenyl)ethylidene]amino}guanidine

  • Cas Number : 792124-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-216852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-216852
Availability details for
Quantity
  • Availability
  • Add to Cart