ENA455586087 Enamine

1-(thiophen-2-ylmethyl)cyclohexane-1-carboxylic acid

  • MDL number MFCD16855601
  • Cas Number : 78502-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-216891
Availability details for
Quantity
  • Availability
  • Add to Cart