ENA455586094 Enamine

ethyl 2-[3-(trifluoromethyl)phenoxy]propanoate

  • MDL number MFCD16516186
  • Cas Number : 1280673-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-216899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-216899
Availability details for
Quantity
  • Availability
  • Add to Cart