ENA455586109 Enamine

2-[(thiophen-2-yl)methyl]bicyclo[2.2.1]heptane-2-carboxylic acid

  • MDL number MFCD16855562
  • Cas Number : 1341128-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-216932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-216932
Availability details for
Quantity
  • Availability
  • Add to Cart