ENA455586152 Enamine

2-[4-(2-oxoethyl)phenyl]acetaldehyde

  • MDL number MFCD17170721
  • Cas Number : 39246-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. -20

Safety Information

msds https://www.enaminestore.com/catalog/EN300-217102
Availability details for
Quantity
  • Availability
  • Add to Cart