ENA455586178 Enamine

5-[(3,4-dimethoxyphenyl)methyl]-5-methyl-1,3-oxazolidin-2-one

  • MDL number MFCD21032407
  • Cas Number : 1498514-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-217197
Availability details for
Quantity
  • Availability
  • Add to Cart