ENA455586348 Enamine

2-(2-methoxyphenyl)pyrimidine-5-carboxylic acid

  • MDL number MFCD17530621
  • Cas Number : 1068977-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-217661
Availability details for
Quantity
  • Availability
  • Add to Cart