ENA455586424 Enamine

N-methoxy-N-methyl-3-(pyrrolidin-1-yl)propanamide

  • MDL number MFCD23725307
  • Cas Number : 1487917-23-6
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-217973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-217973
Availability details for
Quantity
  • Availability
  • Add to Cart