ENA455586446 Enamine

tert-butyl N-methyl-N-[(4-methylbenzenesulfonyl)oxy]carbamate

  • MDL number MFCD24466618
  • Cas Number : 25370-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-218054
Availability details for
Quantity
  • Availability
  • Add to Cart