ENA455586494 Enamine

(2-methylpropyl)(propyl)amine

  • MDL number MFCD09929245
  • Cas Number : 39190-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33331
Availability details for
Quantity
  • Availability
  • Add to Cart