ENA457508147 Enamine

ethyl 2-ethyl-5-methyl-2H-1,2,3-triazole-4-carboxylate

  • MDL number MFCD28954092
  • Cas Number : 942060-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-215851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215851
Availability details for
Quantity
  • Availability
  • Add to Cart