ENA457508174 Enamine

5-(chloromethyl)-4-methyl-2-(propan-2-yl)-1,3-oxazole

  • MDL number MFCD28954119
  • Cas Number : 1803581-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-217182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-217182
Availability details for
Quantity
  • Availability
  • Add to Cart