ENA457508181 Enamine

ethyl 1-ethyl-4-methyl-1H-1,2,3-triazole-5-carboxylate

  • MDL number MFCD28954128
  • Cas Number : 942060-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-217525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-217525
Availability details for
Quantity
  • Availability
  • Add to Cart