ENA457508218 Enamine

2,2-dimethyl-N-[4-(trifluoromethyl)pyridin-2-yl]propanamide

  • MDL number MFCD28954180
  • Cas Number : 1803590-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-218569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218569
Availability details for
Quantity
  • Availability
  • Add to Cart