ENA457508269 Enamine

4-hydroxy-6-methylquinoline-3-carbonitrile

  • MDL number MFCD13181159
  • Cas Number : 61338-30-5
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-181767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181767
Availability details for
Quantity
  • Availability
  • Add to Cart