ENA457508312 Enamine

2-(2-hydroxy-5-nitrophenyl)acetic acid

  • MDL number MFCD04117976
  • Cas Number : 51794-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195992
Availability details for
Quantity
  • Availability
  • Add to Cart