ENA457508318 Enamine

3-[(prop-2-yn-1-yl)carbamoyl]benzoic acid

  • MDL number MFCD12173469
  • Cas Number : 1154949-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-197467
Availability details for
Quantity
  • Availability
  • Add to Cart