ENA457508355 Enamine

ethyl 2-[benzyl(3-ethoxy-3-oxopropyl)amino]propanoate

  • MDL number MFCD16659961

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-204985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204985
Availability details for
Quantity
  • Availability
  • Add to Cart