ENA457508444 Enamine

3-(4-methyl-4H-1,2,4-triazol-3-yl)propan-1-amine dihydrochloride

  • MDL number MFCD28954016
  • Cas Number : 1803585-97-8
  • Molecular Weight : 213.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-112757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112757
Availability details for
Quantity
  • Availability
  • Add to Cart