ENA457508449 Enamine

tert-butyl 2-methyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD20663054
  • Cas Number : 27172-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-210703

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210703
Availability details for
Quantity
  • Availability
  • Add to Cart