ENA457508454 Enamine

dimethyl[(quinolin-2-yl)methyl]amine

  • MDL number MFCD13249726
  • Cas Number : 776-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-211126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211126
Availability details for
Quantity
  • Availability
  • Add to Cart