ENA457508497 Enamine

methyl 3-(hydroxymethyl)-5-nitrobenzoate

  • MDL number MFCD08275104
  • Cas Number : 53732-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-213368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213368
Availability details for
Quantity
  • Availability
  • Add to Cart