ENA457508519 Enamine

(1-benzylazepan-3-yl)methanamine

  • MDL number MFCD22395760
  • Cas Number : 1780225-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-214312
Availability details for
Quantity
  • Availability
  • Add to Cart