ENA457508529 Enamine

2-amino-5-(benzylsulfanyl)benzamide

  • MDL number MFCD13436528
  • Cas Number : 1304873-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-214707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214707
Availability details for
Quantity
  • Availability
  • Add to Cart