ENA457508538 Enamine

1-phenyl-1H-[1,2,3]triazolo[4,5-c]pyridine

  • MDL number MFCD01106350
  • Cas Number : 129303-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-215088
Availability details for
Quantity
  • Availability
  • Add to Cart