ENA457508702 Enamine

2-bromo-1-[2-(trifluoromethyl)pyridin-3-yl]ethan-1-one

  • MDL number MFCD11847723
  • Cas Number : 1379300-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-217763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-217763
Availability details for
Quantity
  • Availability
  • Add to Cart