ENA457508726 Enamine

4-bromo-1-[(3-chlorophenyl)methoxy]-2-methylbenzene

  • MDL number MFCD18978213
  • Cas Number : 854259-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-217950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-217950
Availability details for
Quantity
  • Availability
  • Add to Cart