ENA457508738 Enamine

1-(2,3-dihydro-1,4-benzodioxin-2-yl)-2-(pyridin-2-yl)ethan-1-amine

  • MDL number MFCD21548736
  • Cas Number : 1494421-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-218011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218011
Availability details for
Quantity
  • Availability
  • Add to Cart