ENA457508744 Enamine

2-amino-3,5-dichlorobenzaldehyde

  • MDL number MFCD06657550
  • Cas Number : 53874-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-218029
Availability details for
Quantity
  • Availability
  • Add to Cart