ENA457508761 Enamine

4-methoxy-3-methylbutan-2-amine

  • MDL number MFCD19654937
  • Cas Number : 1341304-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-218138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218138
Availability details for
Quantity
  • Availability
  • Add to Cart