ENA457508774 Enamine

3-amino-5,7-dimethyl-2H,4H,5H,6H,7H-pyrazolo[3,4-d]pyrimidine-4,6-dione

  • MDL number MFCD20698289
  • Cas Number : 67304-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-218170
Availability details for
Quantity
  • Availability
  • Add to Cart