ENA457508792 Enamine

1-{[(tert-butoxy)carbonyl](cyclopropyl)amino}cyclobutane-1-carboxylic acid

  • MDL number MFCD19065414
  • Cas Number : 1490273-29-4
  • Molecular Weight : 255.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-218253

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218253
Availability details for
Quantity
  • Availability
  • Add to Cart