ENA457508865 Enamine

2-(ethylsulfanyl)-6-fluoroaniline

  • MDL number MFCD12149703
  • Cas Number : 1182828-59-6
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-218455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218455
Availability details for
Quantity
  • Availability
  • Add to Cart