ENA457508871 Enamine

4-methyl-1H-pyrazole-3,5-diamine hydrochloride

  • MDL number MFCD28954169
  • Molecular Weight : 148.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-218473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218473
Availability details for
Quantity
  • Availability
  • Add to Cart