ENA457508922 Enamine

2-[(4-cyano-2-fluorophenyl)(methyl)amino]acetic acid

  • MDL number MFCD12109917
  • Cas Number : 1156886-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-218592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218592
Availability details for
Quantity
  • Availability
  • Add to Cart