ENA457508925 Enamine

1-(2-methylmorpholin-4-yl)propan-2-ol

  • MDL number MFCD12764899
  • Cas Number : 1216099-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-218595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218595
Availability details for
Quantity
  • Availability
  • Add to Cart