ENA457508933 Enamine

2-hydroxy-4-{[(1-methyl-1H-pyrazol-5-yl)methyl]amino}benzoic acid

  • MDL number MFCD08696993
  • Cas Number : 1006963-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-218616
Availability details for
Quantity
  • Availability
  • Add to Cart