ENA457508961 Enamine

3-amino-4-(1H-pyrrol-1-yl)phenol

  • MDL number MFCD16622801
  • Cas Number : 1228182-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-218737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218737
Availability details for
Quantity
  • Availability
  • Add to Cart