ENA457509008 Enamine

(1R)-2-chloro-1-[4-(trifluoromethyl)phenyl]ethan-1-ol

  • MDL number MFCD24346567
  • Cas Number : 1072105-58-8
  • Molecular Weight : 224.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-218855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218855
Availability details for
Quantity
  • Availability
  • Add to Cart