ENA457509270 Enamine

2-chloro-5-(trichloromethoxy)pyridine

  • MDL number MFCD28954295
  • Cas Number : 1221171-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-219876
Availability details for
Quantity
  • Availability
  • Add to Cart