ENA457509275 Enamine

1-(benzylsulfanyl)-2-fluoro-5-methyl-4-nitrobenzene

  • MDL number MFCD28954301
  • Cas Number : 1803595-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-219925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219925
Availability details for
Quantity
  • Availability
  • Add to Cart