ENA457509337 Enamine

1-amino-3-(2-methylphenyl)propan-2-ol hydrochloride

  • MDL number MFCD28954391
  • Cas Number : 104338-71-8
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-220968

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-220968
Availability details for
Quantity
  • Availability
  • Add to Cart