ENA457509383 Enamine

2-[(1-phenylcyclopropyl)formamido]acetic acid

  • MDL number MFCD11646746
  • Cas Number : 956101-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-219446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219446
Availability details for
Quantity
  • Availability
  • Add to Cart