ENA457509384 Enamine

tert-butyl N-(3-ethylpyridin-2-yl)carbamate

  • MDL number MFCD11976363
  • Cas Number : 149489-03-2
  • Molecular Weight : 222.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-219458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219458
Availability details for
Quantity
  • Availability
  • Add to Cart