ENA457509396 Enamine

6-chloro-N-[3-(methoxymethyl)phenyl]pyrimidin-4-amine

  • MDL number MFCD16190293
  • Cas Number : 1275675-47-2
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-219487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219487
Availability details for
Quantity
  • Availability
  • Add to Cart