ENA457509397 Enamine

4-chloro-3,5-diiodo-2-methylpyridine

  • MDL number MFCD13188496
  • Cas Number : 1216000-60-0
  • Molecular Weight : 379.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-219490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219490
Availability details for
Quantity
  • Availability
  • Add to Cart